Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARCO Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564134
Description
Immunogen sequence: LNLQARLRVL EMYFLNDTLA AEDSPSFSLL QSAHPGEHLA QGASRLQVLQ AQLTWVRV Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 57%.
MARCO, also known as SR-A6 or SCARA2, is a macrophage surface receptor and a member of the class A scavenger receptor family. As a part of the innate antimicrobial immune system, MARCO can bind both Gram-negative and Gram-positive bacteria via an extracellular, C-terminal, scavenger receptor cysteine-rich (SRCR) domain. In addition to short cytoplasmic and transmembrane domains, there is an extracellular spacer domain and a long, extracellular collagenous domain. The protein may form a trimeric molecule by the association of the collagenous domains of three identical polypeptide chains. Due to its expression in tumor infiltrating macrophages, and its role in inhibiting immune responses within tumor environment, MARCO has been indicated as a potential target for cancer immunotherapy.
Specifications
MARCO | |
Polyclonal | |
Unconjugated | |
Marco | |
AI323439; Ly112; macrophage receptor MARCO; Macrophage receptor with collagenous structure; MARCO; SCARA2; Scavenger receptor class A member 2; scavenger receptor class A, member 2; SR-A6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8685 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
0.30 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UEW3 | |
Marco | |
Recombinant protein corresponding to Human MARCO. Recombinant protein control fragment (Product #RP-103527). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction