Learn More
Invitrogen™ Marginal Zone B Cells Monoclonal Antibody (HIS57), eFluor™ 660, eBioscience™
Mouse Monoclonal Antibody
$425.00
Specifications
Antigen | Marginal Zone B Cells |
---|---|
Clone | HIS57 |
Concentration | 0.2 mg/mL |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-9302
![]() |
Invitrogen™
50948982 |
100 μg |
Each of 1 for $425.00
|
|
|||||
Description
Description: The monoclonal antibody HIS57 recognizes an unknown antigen expressed by rat marginal zone B cells. Marginal zone B cells are an anatomical B cell compartment located in the spleen at the area between red and white pulp. Marginal zone B cells serve as a first line of defense against blood-borne antigens in a T cell-independent manner. Marginal zone B cells can also be phenotyped as B cells that are IgM high, IgD low, CD21 high and CD23 low. Applications Reported: This HIS57 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. Applications Tested: This HIS57 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This HIS57 antibody has also been tested by immunohistochemistry of frozen rat tissue and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emis...
Marginal zone B cells are an anatomical B cell compartment located in the spleen at the area between red and white pulp. Marginal zone B cells serve as a first line of defense against blood-borne antigens in a T cell-independent manner. Marginal zone B cells can also be phenotyped as B cells that are IgM high, IgD low, CD21 high and CD23 low.Specifications
Marginal Zone B Cells | |
0.2 mg/mL | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
eFluor 660 | |
Mouse | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Primary | |
Affinity chromatography |
HIS57 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Rat | |
MZ-B; MZB1 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.