Learn More
Marginal Zone B Cells Mouse Anti-Rat, Brilliant Violet 510, Clone: HIS57, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743342
Description
The HIS57 monoclonal antibody reacts with an unknown antigen that is highly expressed by most marginal zone B (MZ-B) cells in the spleen. In contrast, this antigen is weakly expressed, or not expressed at all, by other B-cell subpopulations. Rat MZ-B cells express low levels of CD45R (mAb HIS24) and sIgD and high levels of sIgM. The HIS57 mAb does not stain granulocytes and thymocytes. Immunohistochemical staining of normal spleen sections with HIS57 mAb produced a positive signal in the marginal zone and, to a lesser extent, in B-cell follicles. This marker can be used in combination with CD45R, sIgD, and sIgM t0 identify MZ-B cells in the rat.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
| Marginal Zone B Cells | |
| HIS57 | |
| Brilliant Violet 510 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat Splenic B Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.