Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marginal Zone B Cells Mouse Anti-Rat, Brilliant Violet 605, Clone: HIS57, BD Optibuild™

Catalog No. BDB743343 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743343 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743343 Supplier BD Biosciences Supplier No. 743343
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIS57 monoclonal antibody reacts with an unknown antigen that is highly expressed by most marginal zone B (MZ-B) cells in the spleen. In contrast, this antigen is weakly expressed, or not expressed at all, by other B-cell subpopulations. Rat MZ-B cells express low levels of CD45R (mAb HIS24) and sIgD and high levels of sIgM. The HIS57 mAb does not stain granulocytes and thymocytes. Immunohistochemical staining of normal spleen sections with HIS57 mAb produced a positive signal in the marginal zone and, to a lesser extent, in B-cell follicles. This marker can be used in combination with CD45R, sIgD, and sIgM t0 identify MZ-B cells in the rat.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Marginal Zone B Cells
Classification Monoclonal
Clone HIS57
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Splenic B Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.