Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Marine Science Outfit
SDPSCI_ED

Catalog No. s85002 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S85002 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S85002 Supplier LaMotte™ Supplier No. 590303
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • Individual test modules for seven key salt water test factors
  • Water sampling bottles
  • Data sheets
  • Three handbooks now available as a download: A Laboratory Manual for Marine Science, Limnology and Investigating Water Problems
  • Soft-sided tote

Learn about saline systems and brackish water.

  • Designed for testing and studying saline systems (oceans, bays, salt marshes) and brackish water
  • Colorimetric test factor for pH (two ranges) and titration test factors for dissolved oxygen, hardness, carbon dioxide, alkalinity and salinity
  • Soft-sided tote is made of durable fabric, solid plastic bottom ready for field work; handy side pockets hold thermometer, pens, writing tablets and other tools

Specifications

Grade 5+
Product Type Marine Science Outfit
Includes Individual test modules for seven key salt water test factors, water sampling bottles, data sheets, and three handbooks now available as a download
Material Fabric, Plastic
For Use With (Application) Designed for testing and studying saline systems (oceans, bays, salt marshes) and brackish water
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.