Learn More
Promega MARK1 Kinase Enzyme System, 10microg
Catalog No. PRV4028 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4028 10 μg
1 options
Catalog No. PRV4028 Supplier Promega Supplier No. V4028
Recombinant full-length human MARK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MARK1 is a member of the MARK family and is a serine/threonine-protein kinase that plays a key role in signal transduction.
  • Easily Screen and Profile MARK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C