Learn More
Promega MARK1 Kinase Enzyme System, 1mg
Catalog No. PRV6446 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6446 1 mg
1 options
Catalog No. PRV6446 Supplier Promega Supplier No. V6446
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human MARK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MARK1 is a member of the MARK family and is a serine/threonine-protein kinase that plays a key role in signal transduction.
  • Easily Screen and Profile MARK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C