Learn More
Invitrogen™ MARK1 Polyclonal Antibody

Description
Immunogen sequence: QEEEELKPYT EPDPDFNDTK RIDIMVTMGF ARDEINDALI NQKYDEVMAT YILLGRKPPE FEGGESLSSG NLCQRSRPSS DLNNSTLQSP AHLKVQRSIS ANQKQRRFSD HAGPSIPPAV SYTKRPQANS VESEQK Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 94%.
Specifications
Specifications
| Antigen | MARK1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MARK1 |
| Gene Accession No. | Q9P0L2 |
| Gene Alias | AW491150; B930025N23Rik; ELKL motif serine/threonine-protein kinase 3; ELKL motif serine-threonine protein kinase 3; Emk3; KIAA1477; MAP/microtubule affinity regulating kinase 1; MAP/microtubule affinity-regulating kinase 1; MARK; MARK1; mark1 {ECO:0000312; microtubule affinity regulating kinase 1; mKIAA1477; mPar-1c; PAR1 homolog c; Par1c; Par-1c; RGD:619882}; serine/threonine-protein kinase MARK1 |
| Gene Symbols | MARK1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.