Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARK1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA517058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517058 Supplier Thermo Scientific Supplier No. PA517058

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

MARK1 (MARK) is a serine/threonine kinase which is involved in cytoskeletal organization and biogenesis. Involved in cell polarity by phosphorylating the microtubule-associated proteins MAP2, MAP4 and MAPT/TAU at KXGS motifs, causing detachment from microtubules, and their disassembly. MARK1 is involved in the regulation of neuronal migration through its dual activities in regulating cellular polarity and microtubule dynamics, possibly by phosphorylating and regulating DCX. Also acts as a positive regulator of the Wnt signaling pathway, probably by mediating phosphorylation of dishevelled proteins (DVL1, DVL2 and/or DVL3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Gene MARK1
Gene Accession No. O08678, Q8VHJ5, Q9P0L2
Gene Alias AW491150; B930025N23Rik; ELKL motif serine/threonine-protein kinase 3; ELKL motif serine-threonine protein kinase 3; Emk3; KIAA1477; MAP/microtubule affinity regulating kinase 1; MAP/microtubule affinity-regulating kinase 1; MARK; MARK1; mark1 {ECO:0000312; microtubule affinity regulating kinase 1; mKIAA1477; mPar-1c; PAR1 homolog c; Par1c; Par-1c; RGD:619882}; serine/threonine-protein kinase MARK1
Gene Symbols MARK1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Ala547 of MARK1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117016, 226778, 4139
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.