Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARK2 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171890R
Description
MARK2 Polyclonal antibody specifically detects MARK2 in Human, Mouse samples. It is validated for Western BlotSpecifications
MARK2 | |
Polyclonal | |
Western Blot | |
EC 2.7.11, EC 2.7.11.1, ELKL motif kinase, ELKL motif kinase 1, EMK-1, EMK1PAR-1, MAP/microtubule affinity-regulating kinase 2MGC99619, PAR1 homolog, Par1b, Ser/Thr protein kinase PAR-1B, serine/threonine protein kinase EMK, serine/threonine-protein kinase MARK2 | |
The immunogen for this product maps to a region between residue 350 and 400 of human MAP/Microtubule Affinity-Regulating Kinase 2 using the numbering given in entry NP_001034558.2 (GeneID 2011). | |
0.1 mL | |
Phospho Specific, Protein Kinase | |
2011 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction