Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mark3 Antibody (SR2263), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322004100UL
Description
Mark3 Monoclonal antibody specifically detects Mark3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
Mark3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR2263 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:30 | |
Cdc25C-associated protein kinase 1, cTAK1, C-TAK1, CTAK1ELKL motif kinase 2, EC 2.7.11, EC 2.7.11.1, EMK2, EMK-2, KP78, MAP/microtubule affinity-regulating kinase 3, PAR1A, Protein kinase STK10, Ser/Thr protein kinase PAR-1, Serine/threonine-protein kinase p78 | |
A synthesized peptide derived from human Mark3 (Uniprot #: P27448) | |
100 μg | |
Protein Kinase | |
4140 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction