Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARK3 Mouse anti-Human, Mouse, Clone:2G12, Abnova™

Catalog No. 89334603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-603 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-603 Supplier Abnova Corporation Supplier No. MAB17748
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human MARK3.

Specifications

Antigen MARK3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2G12
Conjugate Unconjugated
Description MAP/microtubule affinity-regulating kinase 3
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry
Immunohistochemistry (1:200-1:1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene MARK3
Gene Alias CTAK1, KP78, PAR1A
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 435-658 of human MARK3 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4140
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.