Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARK4 Antibody (OTI9B7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272591F
Description
MARK4 Monoclonal specifically detects MARK4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
MARK4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MARK4 | |
Recombinant protein fragment corresponding to amino acids 390-467 of human MARK4 (NP_113605) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI9B7 | |
Immunohistochemistry | |
EC 2.7.11, EC 2.7.11.1, FLJ12177, FLJ90097, KIAA1860MARK4L, MAP/microtubule affinity-regulating kinase 4, MAP/microtubule affinity-regulating kinase like 1, MAP/microtubule affinity-regulating kinase-like 1, MARK4 serine/threonine protein kinase, MARKL1L, MARKL1MARK4S, Nbla00650 | |
Mouse | |
Affinity Purified | |
Apoptosis | |
57787.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction