Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518025
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518025 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518025 Supplier Invitrogen™ Supplier No. PA518025
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

MARK4 is a serine/threonine kinase that is likely to be directly involved in microtubule organization in neuronal cells. The encoded protein is associated with the centrosome throughout mitosis and may be involved in cell cycle control. Expression of MARK4 is a potential marker for cancer, and the encoded protein may also play a role in Alzheimer's disease. Pseudogenes of MARK4 are located on both the short and long arm of chromosome 3. Alternatively spliced transcript variants encoding multiple isoforms have been observed for MARK4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARK4
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Mark4
Gene Accession No. Q8CIP4, Q96L34
Gene Alias 2410090P21Rik; C79806; FLJ90097; KIAA1860; MAP/microtubule affinity regulating kinase 4; MAP/microtubule affinity-regulating kinase 4; MAP/microtubule affinity-regulating kinase 4L; MAP/microtubule affinity-regulating kinase like 1; MAP/microtubule affinity-regulating kinase-like 1; Mark4; MARK4 serine/threonine protein kinase; MARK4L; MARK4S; MARKL1; MARKL1L; microtubule affinity regulating kinase 4; Nbla00650; PAR-1D
Gene Symbols Mark4
Host Species Goat
Immunogen Synthetic peptide sequence (IKRKPPPGCSDSPGV) corresponding to the C-terminus amino acids of MARK4 (aa 674-688).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232944, 57787
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.