Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582137
Description
Immunogen sequence: FVLQDTVEQL RCEHCARFLA DRFVEGVCPF CGYEEARGDQ CDKCGKLINA VELKKPQCKV CRSCPVVQSS QHLFLDLPKL EKRLEEWLGR TLPGSDWTPN AQFITRSWLR DGLKPRCITR DLKWGTPVPL EGFEDKVFYV WFD.
Methionyl-tRNA synthetase belongs to the class I family of tRNA synthetases. It catalyzes the chemical reaction:ATP + L-methionine + tRNA(Met) = AMP + diphosphate + L-methionyl-tRNA(Met).
Specifications
MARS | |
Polyclonal | |
Unconjugated | |
MARS1 | |
CMT2U; cytosolic methionyl-tRNA synthetase; id:ibd5106; ILFS2; ILLD; MARS; mars protein; mars1; methionine tRNA ligase; methionine--tRNA ligase, cytoplasmic; methionine-tRNA synthetase; methionyl trna synthetase; methionyl-tRNA synthetase; methionyl-tRNA synthetase 1; methionyl-tRNA synthetase, cytoplasmic; metRS; MRS; Mtrns; SPG70; Unknown (protein for MGC:134096); zgc:66122 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4141 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P56192 | |
MARS1 | |
Recombinant protein corresponding to Human MARS. Recombinant protein control fragment (Product #RP-102035). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction