Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARS Polyclonal Antibody

Catalog No. PIPA5121785
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121785 20 μL
PIPA5119284 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121785 Supplier Invitrogen™ Supplier No. PA5121785
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MARS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Methionyl-tRNA synthetase belongs to the class I family of tRNA synthetases. It catalyzes the chemical reaction:ATP + L-methionine + tRNA(Met) = AMP + diphosphate + L-methionyl-tRNA(Met).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MARS1
Gene Accession No. P56192, Q68FL6
Gene Alias CMT2U; cytosolic methionyl-tRNA synthetase; id:ibd5106; ILFS2; ILLD; MARS; mars protein; mars1; methionine tRNA ligase; methionine--tRNA ligase, cytoplasmic; methionine-tRNA synthetase; methionyl trna synthetase; methionyl-tRNA synthetase; methionyl-tRNA synthetase 1; methionyl-tRNA synthetase, cytoplasmic; metRS; MRS; Mtrns; SPG70; Unknown (protein for MGC:134096); zgc:66122
Gene Symbols MARS1
Host Species Rabbit
Immunogen Recombinant protein Methionyl tRNA Synthetase. The antigen corresponds to amino acid range 74-212 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216443, 4141
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.