Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531082 Supplier Invitrogen™ Supplier No. PA531082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, MCF-7. Predicted reactivity: Mouse (83%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Methionine-tRNA ligase (EC 6.1.1.10) catalyzes the chemical reaction: ATP + L-methionine + tRNAMet <--> AMP + diphosphate + L-methionyl-tRNAMetThe 3 substrates of this enzyme are ATP, L-methionine, and tRNA(Met), whereas its 3 products are AMP, diphosphate, and L-methionyl-tRNA(Met).This enzyme participates in 3 metabolic pathways: methionine metabolism, selenoamino acid metabolism, and aminoacyl-trna biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARS2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MARS2
Gene Accession No. Q96GW9
Gene Alias C730026E21Rik; COXPD25; EC 6.1.1.10; MARS2; methionine tRNA ligase 2, mitochondrial; methionine--tRNA ligase 2; methionine--tRNA ligase, mitochondrial; methionine-tRNA synthetase 2 (mitochondrial); methionine-tRNA synthetase 2, mitochondrial; methionyl-tRNA synthetase 2; methionyl-tRNA synthetase 2, mitochondrial; methionyl-tRNA synthetase, mitochondrial; MetRS; mitochondrial methionine--tRNA ligase; mitochondrial methionyl-tRNA synthetase; mtMetRS; RGD1311527
Gene Symbols MARS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human MARS2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92935
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.