Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MART-1/Melan-A Mouse anti-Human, Alexa Fluor 647, Clone: A103 (also known as A103), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567883
This item is not returnable.
View return policy
Description
MART-1/Melan-A Monoclonal antibody specifically detects MART-1/Melan-A in Human samples. It is validated for Intracellular staining (Flow Cytometry), Immunofluorescence, Immunohistochemistry-formalin (antigen retrieval required)
Specifications
MART-1/Melan-A | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
A103 (also known as A103) | |
Alexa Fluor 647 | |
MART-1; Melan-A; Melan A ; MLANA | |
Recombinant Human Melan-A Protein | |
RUO | |
Human | |
Liquid |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction