Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARVELD2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074633H
Description
MARVELD2 Polyclonal antibody specifically detects MARVELD2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
MARVELD2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
MARVEL domain containing 2, MRVLDC2, TRICautosomal recessive 49 | |
The immunogen recognized by this antibody maps to a region between residue 90 and 140 of human MARVEL domain containing 2 (Tricellulin) using the numbering given in entry NP_001033692.1 (GeneID 153562). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
153562 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction