Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mas Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178444AF350
Description
Mas Polyclonal specifically detects Mas in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Mas | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas | |
Rabbit | |
Affinity Purified | |
Cancer, GPCR, Neuroscience | |
4142.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
MAS1 | |
A synthetic peptide made to an internal portion of the human MAS1 protein (between residues 75-125) [Uniprot: P04201] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Primate, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction