Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mas Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178444AF647
Description
Mas Polyclonal specifically detects Mas in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Mas | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
P04201 | |
MAS1 | |
A synthetic peptide made to an internal portion of the human MAS1 protein (between residues 75-125) [Uniprot: P04201] | |
0.1 mL | |
Cancer, GPCR, Neuroscience | |
4142 | |
Human, Mouse, Rat, Primate, Rabbit | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, monkey, rat, and mouse. Rabbit reactivity reported in the scientific literature (PMID: 23701246). | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction