Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ Omni·CARDIO™ Control

Catalog No. 22640058
Change view
Click to view available options
Level:
1
2
3
Low
Tri-Level Multi-Pack
Ultra Low
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
22-640-058 2
22-640-055 Ultra Low
22-640-056 Low
22-640-057 1
22-640-059 3
22-640-060 Tri-Level Multi-Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-640-058 Supplier Thermo Scientific™ Supplier No. OCRD202

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

This item is not returnable. View return policy

Ensure accurate results in testing of traditional cardiac marker panels and the new generation of STAT analytes including D-Dimer, hCG, Myeloperoxidase and Procalcitonin.

Ensure accurate results in testing of traditional cardiac marker panels as well as the new generation of STAT analytes including D-Dimer, hCG, Myeloperoxidase and Procalcitonin with the Thermo Scientific™ MAS™ Omni·CARDIO™ Control.

  • Matrix: Human Serum
  • Packaging: 6 x 3mL assayed
  • Value assignment provided for key instrument systems
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage
  • 36 month shelf life from date of manufacture when stored at -25°C to -15°C
  • 15 days at 2°C to 8°C (for BNP-32, CK-MB, D-Dimer, Digitoxin, hCG, hsCRP, Myeloperoxidase, Procalcitonin, Total CK, Troponin-I and Troponin-T)
  • 10 days at 2°C to 8°C (for Myoglobin and NT-proBNP)
Detectable Analytes β-hCG, Brain Natriuretic Peptide-32 (BNP-32), Creatinine Kinase-MB (CK-MB), D-dimer, Digitoxin, High Sensitivity C-Reactive Protein (hsCRP), hCG, Myeloperoxidase (MPO), Myoglobin, N-Terminal Brain Natriuretic Peptide (NT-proBNP), Procalcitonin (PCT), Total Creatinine Kinase (Total CK), Troponin I, Troponin T
Sample Type Serum
Type Control
Level 2
Packaging 6
Quantity 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.