Learn More
Thermo Scientific™ MAS™ Omni·CARDIO™ Control
Description
Ensure accurate results in testing of traditional cardiac marker panels as well as the new generation of STAT analytes including D-Dimer, hCG, Myeloperoxidase and Procalcitonin with the Thermo Scientific™ MAS™ Omni·CARDIO™ Control.
- Matrix: Human Serum
- Packaging: 6 x 3mL assayed
- Value assignment provided for key instrument systems
Specifications
Specifications
| Content And Storage |
|
| Detectable Analytes | β-hCG, Brain Natriuretic Peptide-32 (BNP-32), Creatinine Kinase-MB (CK-MB), D-dimer, Digitoxin, High Sensitivity C-Reactive Protein (hsCRP), hCG, Myeloperoxidase (MPO), Myoglobin, N-Terminal Brain Natriuretic Peptide (NT-proBNP), Procalcitonin (PCT), Total Creatinine Kinase (Total CK), Troponin I, Troponin T |
| Sample Type | Serum |
| Type | Control |
| Level | 3 |
| Packaging | 6 |
| Quantity | 3 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.