Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ Omni·CORE Controls

Catalog No. p-4527742
Change view
Click to view available options
Level:
1
2
3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
22-640-051 1
22-640-052 2
22-640-053 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-640-051 Supplier Thermo Scientific™ Supplier No. OCR101

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

This item is not returnable. View return policy

Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels.

Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo Scientific™ MAS™ Omni·CORE Controls. Value assignment is provided for key integrated instrument systems.

  • Three distinct levels
  • Liquid, ready-to-use
  • Human serum based matrix
  • Assayed values available for most analyzers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 36 month shelf life from date of manufacture when stored at -25° to -15°C; 30-day open-vial stability when stored at 2° to 8°C
Description Omni CORE Control, Assayed
Detectable Analytes Acetaminophen, Albumin, Alkaline Phosphatase, alpha-1-Acid Glycoprotein, alpha-1-Antitrypsin, alpha-2-Macroglobulin, ALT, Amikacin, Amylase, Amylase (Pancreatic), Antistreptolysin O (ASO), Apolipoprotein A1, Apolipoprotein B, AST, beta-2-microglobulin (B2M), Bile Acids, Bilirubin Conjugated, Bilirubin Direct (DBIL), Bilirubin Neonatal, Bilirubin Total (TBIL), Bilirubin Unconjugated, BUN, C3 Complement, C4 Complement, Caffeine, Calcium, Carbamazepine, Ceruloplasmin, Chloride, Cholesterol, Creatine Kinase (CK), CO2, Copper, Cortisol, C-Reactive Protein (CRP), Creatinine, Digoxin, Disopyramide, Ethanol, Ethosuximide, Ferritin, Gentamicin, GGT, Glucose, Glutamate Dehydrogenase (GLDH), Haptoglobin, HDL, IgA, IgE, IgG, IgM, Iron, Iron Binding Capacity (IBC), Iron Binding Capacity, Total (TIBC), Iron Binding Capacity, Unsaturated (UIBC), Lactic Acid, LDH, LDL, Lidocaine, Lipase, Lipoprotein (a), Lithium, Magnesium, Methotrexate, N-acetylprocainamide (NAPA), Osmolality, Phenobarbital, Phenytoin, Phospholipids, Phosphorus, Potassium, Prealbumin, Primidone, Procainamide, Protein Electrophoresis, Pseudocholinesterase (PCHE), Quinidine, Rheumatoid Factor, Salicylate (SALY), Sodium, Thyroxine (T4 Total), Theophylline, Tobramycin, Total Protein, Transferrin (TRF), Tricyclic Antidepressants, Triglycerides, Thyroid Stimulating Hormone (TSH), T-uptake, Uric Acid, Valproic Acid, Vancomycin, Zinc
Sample Type Serum
Type Control
Level 1
Packaging 6 Pk.
Quantity 5 mL
Stability 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.