Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ Omni CORE™ Max Level 1

Catalog No. 22048138
Click to view available options
Level:
1
This item is not returnable. View return policy
This item is not returnable. View return policy

MAS™ Omni CORE Max consolidates general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo Scientific™ MAS™ Omni CORE Controls.

  • Value assignment is provided for key integrated instrument systems.
  • Liquid ready-to-use
  • Human-serum based matrix
  • Assayed values available for most analyzers
  • Load-and-Go barcoded¹
  • Tubes for improved efficiency and daily time savings

Need to add: ¹Barcodes available for selected instruments. Availability of products in each country depends on local regulatory marketing authorization status.

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Human Serum
Type Chemistry Control
Disposable Reusable
Level 1
Sterility Non-sterile
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.