Learn More
Thermo Scientific™ MAS™ Omni·IMMUNE™ PRO Immunoassay Controls

Description
Consolidate your immunoassay and tumor marker controls into a single QC product with Thermo Scientific™ MAS™ Omni·IMMUNE™ PRO Immunoassay Controls. This QC product contains the same analyte panel as the Omni·IMMUNE control but also has the following additional analytes: Anti-Thyroglobulin (Anti-Tg), Anti-Thyroid Peroxidase (Anti-TPO) and Sex Hormone Binding Globulin (SHBG).
- Liquid, ready-to-use
- Human-based serum matrix
- Value assignment provided for most major analyzers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | -25°C to -15°C |
| Description | Omni IMMUNE PRO Control, Assayed |
| Detectable Analytes | 17-OH-Progesterone, 25-OH-Vitamin D, Acetaminophen, Adrenocorticotropic Hormone (ACTH), Aldosterone, alpha-Fetoprotein (AFP), Amikacin, Anti-Tg, Anti-TPO, Benzodiazepines, beta-2-microglobulin (B2M), β-hCG, CA-125, CA 15-3, CA 19-9, Carbamazepine, Carcinoembryonic Antigen (CEA), Cortisol, C-peptide, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Estriol (Free), Ethosuximide, Ferritin, Folate (Vitamin B9), Fructosamine, Follicle Stimulating Hormone (FSH), Gastrin, Gentamicin, Growth Hormone, hCG, Homocysteine, IgE, Insulin-like Growth Factor 1 (IGF-1), Insulin, Luteinizing Hormone (LH), Lidocaine, N-acetylprocainamide (NAPA), Prostatic Acid Phosphatase (PAP), Phenobarbital, Phenytoin, Phenytoin (Free), Primidone, Procainamide, Procalcitonin (PCT), Progesterone, Prolactin, PSA (Free), PSA (Total), Parathyroid Hormone (PTH), Quinidine, Salicylate (SALY), Sex Hormone-binding Globulin (SHBG), T3 (Free), Triiodothyroxine (T3 Total), T4 (Free), Thyroxine (T4 Total), Thyroxine-binding Globulin (TBG), Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, Thyroid Stimulating Hormone (TSH), T-uptake, Valproic Acid, Valproic Acid (Free), Vancomycin, Vitamin B12 |
| Sample Type | Serum |
| Type | Control |
| Level | 2 |
| Packaging | 6 Pk. |
| Quantity | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.