Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAS™ Omni·IMMUNE™ PRO™ Max Level 2
Click to view available options
Sample Type:
Human Serum
Description
MAS Omni·CORE Max consolidates general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo Scientific™ MAS™ Omni·CORE Controls. Value assignment is provided for key integrated instrument systems.
- Three distinct levels
- liquid ready-to-use
- Human-serum based matrix
- Assayed values available for most analyzers
- Load-and-Go barcoded¹ tubes for improved efficiency and daily time savings
Required addition:
- ¹Barcodes available for selected instruments.
- Availability of products in each country depends on local regulatory marketing authorization status.

Specifications
Specifications
Sample Type | Human Serum |
Type | Clinical Chemistry Control |
Disposable | Reusable |
Level | 2 |
Sterility | Non-sterile |
Form | Liquid Ready-to-Use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction