Learn More
Thermo Scientific™ MAS™ Omni·Infectious™ Quality Controls
Description
Assess the performance of Serological assays for viral and bacterial pathogens using the Thermo Scientific MAS Omni Infectious Quality Controls.
Assess the performance of serological assays for viral blood stream infections using the Thermo Scientific MAS Omni Infectious Quality Controls. These third-party control panels include individual positive controls for anti-HBs, HIV-p24 and Syphilis as well as consolidated HIV, HCV, HBV and HTLV for efficiency.
- 30 days open vial stability for Syphilis; anti-HBs and HIV p24 at 2 to 8°C
- Ready to use straight from the refrigerator
- Human serum matrix
- BSI Positive and Infectious Negative
- BSI Positive has Multiple analytes in a single vial
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | MAS Syphilis Positive Control |
| Detectable Analytes | anti-TP (syphilis), HIV, HBV, HTLV |
| Type | Control |
| Packaging | One box of control contains five tubes of Syphilis Positive Control |
| Quantity | 5 x 1.0 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.