Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ SARS-CoV-2 Serology Quality Controls

Catalog No. p-9255002
Click to view available options
Type:
SARS-CoV-2 IgG Positive Control
SARS-CoV-2 Negative Control
This item is not returnable. View return policy

Catalog No. 10-028-305

Supplier: Thermo Scientific™ 10028305

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Assess the performance of serological assays for SARS-CoV-2 infections using the Thermo Scientific MAS SARS-CoV-2 Serology Quality Controls. These third-party controls include positive controls for SARS-CoV-2 and a negative control for accuracy.

Assess the performance of serological assays for SARS-CoV-2 infections using the Thermo Scientific MAS SARS-CoV-2 Serology Quality Controls. These third-party controls include individual positive control for SARS-CoV-2 as well as a negative control for accuracy.

  • 21 days open vial stability at 2°C to 8°C
  • Ready to use straight from the refrigerator
  • Human Plasma matrix
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes SARS-CoV-2 Virus IgG Ab
Type SARS-CoV-2 IgG Positive Control
Packaging One box of control contains five tubes SARS-CoV-2 IgG Positive Control
Quantity 5 x 3.0 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.