Learn More
Thermo Scientific™ MAS™ T-Marker™ Controls
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte Thermo Scientific™ MAS™ T-Marker Controls.
Supplier:  Thermo Scientific™ TUM101
 View more versions of this product
Catalog No. TU1101
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte Thermo Scientific™ MAS™ T-Marker Controls. These controls contain more 24 analytes, including a complete offering of cancer-associated antigens.
  • Liquid, ready to use; no additional preparation required
  • Human serum matrix
  • Includes CA 15-3, CA 19-9, CA 72-4, CA-125, CA 27-29
  • Assayed values available for most major analyzers.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.