Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAS™ UrichemTRAK Controls
GSA_VA

Catalog No. 23139109
Change view
Click to view available options
Description:
UrichemTRAK Controls, Assayed
UrichemTRAK Controls, Assayed, Bi-level
Level:
1
2
Bi-level
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Level
23-139-109 UrichemTRAK Controls, Assayed 1
23-139108 UrichemTRAK Controls, Assayed, Bi-level Bi-level
23-139110 UrichemTRAK Controls, Assayed 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-139-109 Supplier Thermo Scientific™ Supplier No. UR11001

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

This item is not returnable. View return policy

Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient vial format.

Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient Vista™ vial format. Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.

  • Liquid, human urine matrix
  • Open and use; no equilibration required
  • Two-year open-vial stability
  • Individual levels or multi-pack configurations
  • Stable for 24 months at 2° to 8°C
  • Assayed values available for most major analyzers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description UrichemTRAK Controls, Assayed
Detectable Analytes Amylase, Calcium, Chloride, Cortisol, Creatinine, Glucose, hCG, Magnesium, Microalbumin, Osmolality, pH, Phosphorus, Potassium, Sodium, Specific Gravity, Total Protein, Urea Nitrogen, Uric Acid
Sample Type Urine
Type Control
Level 1
Packaging 6 pk.
Quantity 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.