Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533918
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533918 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533918 Supplier Invitrogen™ Supplier No. PA533918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Mouse, Rat, Bovine, Dolphin, Hamster, Elephant, Panda, Pig, Opossum (100%) Platypus (94%).

The structure of the MAS1 product indicates that it belongs to the class of receptors that are coupled to GTP-binding proteins and share a conserved structural motif, which is described as a 7-transmembrane segment following the prediction that these hydrophobic segments form membrane-spanning alpha-helices. The MAS1 protein may be a receptor that, when activated, modulates a critical component in a growth-regulating pathway to bring about oncogenic effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MAS1
Gene Accession No. P04201, P12526, P30554
Gene Alias angiotensin-(1-7) receptor; c-mas; MAS; MAS proto-oncogene; MAS1; Mas-1; MAS1 oncogene; Mas1 proto-oncogene; MAS1 proto-oncogene, G protein-coupled receptor; Mas-related G protein-coupled receptor a; MGC119966; MGRA; Proto-oncogene Mas
Gene Symbols MAS1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human MAS1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100522470, 17171, 25153, 4142, 783010
Target Species Human, Mouse, Rat, Monkey, Bovine, Hamster, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.