Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543668 Supplier Invitrogen™ Supplier No. PA543668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RPKYQSALVC ALLWALSCLV TTMEYVMCID REEESHSRND CRAVIIFIAI Sequence homology: Guinea Pig: 77%; Human: 100%; Mouse: 77%; Rat: 77%.

The structure of the MAS1 product indicates that it belongs to the class of receptors that are coupled to GTP-binding proteins and share a conserved structural motif, which is described as a 7-transmembrane segment following the prediction that these hydrophobic segments form membrane-spanning alpha-helices. The MAS1 protein may be a receptor that, when activated, modulates a critical component in a growth-regulating pathway to bring about oncogenic effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAS1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAS1
Gene Accession No. P04201
Gene Alias angiotensin-(1-7) receptor; c-mas; MAS; MAS proto-oncogene; MAS1; Mas-1; MAS1 oncogene; Mas1 proto-oncogene; MAS1 proto-oncogene, G protein-coupled receptor; Mas-related G protein-coupled receptor a; MGC119966; MGRA; Proto-oncogene Mas
Gene Symbols MAS1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MAS1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4142
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.