Learn More
MAS1 Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier: Thermo Scientific PA5111828
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)KIRKNTW ASHSSK Homology: Mouse, human amino acid residues are identical.
The structure of the MAS1 product indicates that it belongs to the class of receptors that are coupled to GTP-binding proteins and share a conserved structural motif, which is described as a 7-transmembrane segment following the prediction that these hydrophobic segments form membrane-spanning alpha-helices. The MAS1 protein may be a receptor that, when activated, modulates a critical component in a growth-regulating pathway to bring about oncogenic effects.
Specifications
| MAS1 | |
| Polyclonal | |
| Unconjugated | |
| MAS1 | |
| c-mas; Mas; Mas1; Mas-1; Mas1 proto-oncogene; MGC119966; MGRA; Proto-oncogene Mas | |
| Guinea Pig | |
| Antigen affinity chromatography | |
| RUO | |
| 17171, 25153, 4142 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P04201, P12526, P30554 | |
| MAS1 | |
| Peptide corresponding to amino acid residues 212-224 of the rat Angiotensin-(1-7) Mas receptor (3rdintracellularloop) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.