Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAS1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109942 Supplier Invitrogen™ Supplier No. PA5109942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVWGKICWFS QRAGWTVFAE SQISLSCSLC LHSGDQEAQN PNLVSQLCGV FLQNETNETI HMQMSMAVGQ.

The proto-oncogene MAS1 is a G protein-coupled receptor located on the plasma membrane. In transfected NIH/3T3 cells, MAS1 has a weak focus-inducing activity. MAS1 is an antagonist of the AT1 receptor (Angiotensin II type 1 receptor), inhibiting the actions of Angiotensin II. MAS1L (MAS1-like), also known as MAS-L or MRG (Mas-related G-protein coupled receptor MRG), is a 378 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 (GPCR) family. MAS1 related proteins are thought to regulate nociceptor function and development, including the sensation or modulation of pain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAS1L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MAS1L
Gene Accession No. Q7TN38
Gene Alias DAAP-341G9.4; dJ994E9.2; MAS1 oncogene-like; MAS1 proto-oncogene like, G protein-coupled receptor; MAS1L; MAS1-like; MAS-L; MAS-R; mas-related G protein-coupled MRG; Mas-related G protein-coupled receptor b; MAS-related gene H; MAS-related GPR, member H; mas-related G-protein coupled receptor member H; Mas-related G-protein coupled receptor MRG; MAS-related G-protein coupled receptor, member H; MGC119987; Mgrb; MRG; Mrg and gpcr; Mrgh; MrgHGPR90; Mrgprh
Gene Symbols MAS1L
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human MAS1L (NP_443199.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404641
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.