Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAS1L Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA120468

Catalog No. PIPA120468


Description

Description

PA1-20468 detects G Protein-Coupled Receptor MRG in Human samples.

The proto-oncogene MAS1 is a G protein-coupled receptor located on the plasma membrane. In transfected NIH/3T3 cells, MAS1 has a weak focus-inducing activity. MAS1 is an antagonist of the AT1 receptor (Angiotensin II type 1 receptor), inhibiting the actions of Angiotensin II. MAS1L (MAS1-like), also known as MAS-L or MRG (Mas-related G-protein coupled receptor MRG), is a 378 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 (GPCR) family. MAS1 related proteins are thought to regulate nociceptor function and development, including the sensation or modulation of pain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAS1L
Polyclonal
Unconjugated
MAS1L
DAAP-341G9.4; dJ994E9.2; MAS1 oncogene-like; MAS1 proto-oncogene like, G protein-coupled receptor; MAS1L; MAS1-like; MAS-L; MAS-R; mas-related G protein-coupled MRG; Mas-related G protein-coupled receptor b; MAS-related gene H; MAS-related GPR, member H; mas-related G-protein coupled receptor member H; Mas-related G-protein coupled receptor MRG; MAS-related G-protein coupled receptor, member H; MGC119987; Mgrb; MRG; Mrg and gpcr; Mrgh; MrgHGPR90; Mrgprh
Rabbit
Antigen affinity chromatography
RUO
116511
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
P35410
MAS1L
Synthetic 20 amino acid peptide from C-terminus of human MAS1L
25 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.