Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAS1L Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS5926R

Description
The proto-oncogene MAS1 is a G protein-coupled receptor located on the plasma membrane. In transfected NIH/3T3 cells, MAS1 has a weak focus-inducing activity. MAS1 is an antagonist of the AT1 receptor (Angiotensin II type 1 receptor), inhibiting the actions of Angiotensin II. MAS1L (MAS1-like), also known as MAS-L or MRG (Mas-related G-protein coupled receptor MRG), is a 378 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 (GPCR) family. MAS1 related proteins are thought to regulate nociceptor function and development, including the sensation or modulation of pain.Specifications
MAS1L | |
Polyclonal | |
Unconjugated | |
MAS1L | |
DAAP-341G9.4; dJ994E9.2; MAS1 oncogene-like; MAS1 proto-oncogene like, G protein-coupled receptor; MAS1L; MAS1-like; MAS-L; MAS-R; mas-related G protein-coupled MRG; Mas-related G protein-coupled receptor b; MAS-related gene H; MAS-related GPR, member H; Mas-related G-protein coupled receptor member H; Mas-related G-protein coupled receptor MRG; MAS-related G-protein coupled receptor, member H; MGC119987; Mgrb; MRG; Mrg and gpcr; Mrgh; MrgHGPR90; Mrgprh | |
Rabbit | |
Protein A | |
RUO | |
116511 | |
-20°C | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P35410 | |
MAS1L | |
KLH conjugated synthetic peptide derived from human MAS1L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction