Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mash1/Ascl1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991577
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1577 100 μg
50-199-1578 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1577 Supplier Rockland Immunochemicals Supplier No. 600101MH4
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse Mash1/Ascl1. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse and rat, and 93% homology with human.

ASCL1 (also known as ASH1) is a basic helix-loop-helix transcription factor that is required for early development of the nervous system. Expressed in fetal brain, ASCL1 is essential for the proper development of autonomic neurons and for the survival of subsets of autonomic neurons. ASCL1 interaction with MEF-2A may regulate the expression of specific genes that are critical for the formation of distinct neuronal circuits within the central nervous system. The high level of ASCL1 expression in neuroendocrine tumors, such as medullary thyroid cancer, small cell lung cancer and lung cancer with neuroendocrine features may provide a useful marker for cancers with neuroendocrine features. Mapping to human chromosome 12, the ASCL1 gene contains a trinucleotide repeat region, making this locus a candidate for inherited disease.

Specifications

Antigen Mash1/Ascl1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Ascl1
Gene Accession No. P50553, Q02067
Gene Alias achaete scute protein; achaete-scute complex homolog 1; achaete-scute complex homolog 1 (Drosophila); achaete-scute complex homolog-like 1; achaete-scute complex-like 1; achaete-scute complex-like protein 1; achaete-scute family bHLH transcription factor 1; Achaete-scute homolog 1; achaete-scute homolog 1-like; AI225900; ASCL1; ASH1; ASH-1; BHLHA46; Class A basic helix-loop-helix protein 46; HASH1; KMT2H; LOC100342114; mammalian achaete scute homolog 1; Mash1; Mash-1
Gene Symbols Ascl1
Host Species Goat
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17172, 429
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.