Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Masitinib
GREENER_CHOICE

Catalog No. 770510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-051-0 10 mg
77-055-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 77-051-0 Supplier Tocris Bioscience™ Supplier No. 7705/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent colony-stimulating factor 1 receptor (CSF-1R) tyroine kinase inhibitor

Masitinib is a potent inhibitor of the tyrosine kinase receptor colony-stimulating factor 1R (CSF-1R) (IC50 = 90 nM). It is also an NLRP3 inflammasome activator. Masitinib additionally inhibits the proteolytic activity of SARS-CoV-2 main protease (Mpro) and blocks viral replication, as well as acting as a PDGFRα/β inhibitor. In vivo, Masitinib ameliorates cognitive impairment in transgenic mice model of Alzheimer's disease, and slows disease progression in a rat model of amyotrophic lateral sclerosis (ALS). Orally bioavailable.

Specifications

Chemical Name or Material 4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-thiazolyl]amino]phenyl]benzamide
CAS 790299-79-5
Quantity 10 mg
Target CD117/c-kit
Molecular Formula C28H30N6OS
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.