Learn More
3M™ Masking Tape 2307
Catalog No. 19072024 Shop All 3M Products
Change view
Click to view available options
Quantity:
24 Rolls/Cs.
36 Rolls/Cs.
Dimensions (L x W):
55 m x 24 mm
55 x 36 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dimensions (L x W)
19-072-024 36 Rolls/Cs. 55 m x 24 mm
19-072-119 24 Rolls/Cs. 55 x 36 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-024 Supplier 3M™ Supplier No. 7000123531
Add to Cart
Add to Cart
3M™ Masking Tape 2307 was formulated for general use, with a broad tack that will adhere to a variety of surfaces. The rubber adhesive sticks instantly with moderate strength. The crepe paper backing conforms over irregular surfaces and around corners while holding reliably. It is especially useful in woodworking and cabinet making applications.
  • Easily identifiable, general purpose tape for woodworking
  • Rubber adhesive offers broad tack and moderate adhesion
  • Sticks quickly and holds reliably
  • Crepe-paper backing conforms over irregular surfaces, contours, and around corners
  • Rubber adhesive sticks instantly with moderate strength.
  • Crepe paper backing conforms over irregular surfaces and around corners while holding reliably.
  • Especially useful in woodworking and cabinet making applications.
TRUSTED_SUSTAINABILITY
Specifications
Color Tan
Material Rubber
Product Type Masking Tape
For Use With (Application) Aerospace, Appliance, Building and Construction, Commercial and Specialty Vehicles, Equipment and Facility Maintenance, Exterior and Glass, Heavy Industrial, Interior, Marine, Metalworking, Paper and Print, Personal Recreation Vehicles, Rail, Rig Fabrication and Maintenance, Woodworking
Model 2307
Quantity 36 Rolls/Cs.
Thickness (English) 5.2 mil
Dimensions (L x W) 55 m x 24 mm