Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MASP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596397 Supplier Invitrogen™ Supplier No. PA596397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa Immunogen sequence: HTVELNNMFG QIQSPGYPDS YPSDSEVTWN ITVPDGFRIK LYFMHFNLES SYLCEYDYVK VETEDQVLAT FCGRETTDTE QTPGQEVVLS PGSFMSITFR SDFSNEERFT GFDAHYMAVD VDECKEREDE ELSCDHYCHN YIGGYYCSCR FGYILHTDNR TCRVECSDNL FTQRTGVITS PDFPNPYPKS SECLYTIELE EGFMVNLQFE DIFDIEDHPE VPCPYDYIKI KVGPKVLGPF CGEKAPEPIS TQSHSVLILF HSDNSGENRG WRLSYRAAGN E.

MBL is a key component in immune response in that it can directly trigger neutralization of invading microorganisms by an Ab-independent mechanism. Mutations of human MBL are associated with immunodeficiency resulting from a reduction in the ability of the mutant MBL to initiate complement fixation. In human, three types of MBL-associated serine proteases, MASP-1, MASP-2 and MASP-3, and a truncated form of MASP-2 (small MBL-associated protein; sMAP or MAp19) complex with MBL to activate the lectin pathway of the complement system. MASP-3 is an alternatively spliced product from the MASP-1 gene. The heavy/A chains are identical between MASP-1 and MASP-3 but the light/B chains are entirely different. Activated MASPs subsequently cleave and activate downstream components of the complement pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MASP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene MASP1
Gene Accession No. P48740, P98064, Q8CHN8
Gene Alias 3MC1; AW048060; C4/C2 activating component of Ra-reactive factor; CCPII; Complement factor MASP-3; complement-activating component of Ra-reactive factor; Crarf; CRARF1; mannan binding lectin serine peptidase 1; mannan-binding; mannan-binding lectin serine peptidase 1; mannan-binding lectin serine peptidase 1 (C4/C2 activating component of Ra-reactive factor); mannan-binding lectin serine protease 1; Mannan-binding lectin serine protease 1 heavy chain; Mannan-binding lectin serine protease 1 light chain; Mannose-binding lectin-associated serine protease 1; mannose-binding protein associated serine protease-1; mannose-binding protein-associated serine protease; MAP1; MAp44; MASP; MASP1; MASP-1; Masp1/3; MASP3; PRSS5; Ra-reactive factor serine protease p100; raRF; Serine protease 5; serine protease MASP1
Gene Symbols MASP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 20-300 of human MASP1 (NP_0018703).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17174, 5648, 64023
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.