Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MASP2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299162PCP
Description
MASP2 Polyclonal antibody specifically detects MASP2 in Human samples. It is validated for Western BlotSpecifications
MASP2 | |
Polyclonal | |
Western Blot | |
EC 3.4.21, EC 3.4.21.104, mannan-binding lectin serine peptidase 1 pseudogene 1, mannan-binding lectin serine peptidase 2, mannan-binding lectin serine protease 1 pseudogene 1, mannan-binding lectin serine protease 2, Mannose-binding protein-associated serine protease 2, MAP19, MASP1P1, MASP-2, MBL-associated plasma protein of 19 kD, MBL-associated serine protease 2, small MBL-associated protein, sMAP | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MASP2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
10747 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction