Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MASP2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA556111
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556111 Supplier Thermo Scientific Supplier No. PA556111
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVCEPVCGLS ARTTGGRIYG GQKAKPGDFP WQVLILGGTT AAGALLYDNW VLTAAHAVYE QKHDASALDI RMGTLKRLSP HYTQAWSEAV FIHEGYTHDA GFDNDIALIK LNNKVVINSN ITPICLPRKE AESFMRTDDI GTASGWGL Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

Mannose (or mannan)-binding lectin (MBL), also known as serum mannosebinding protein (MBP), initiates the lectin branch of the innate immune response by binding to the surface of potentially pathogenic microorganisms and initiating complement fixation through an N-terminal collagen-like domain. MBL is a key component in immune response in that it can directly trigger neutralization of invading microorganisms by an Ab-independent mechanism. Mutations of human MBL are associated with immunodeficiency resulting from a reduction in the ability of the mutant MBL to initiate complement fixation. In human, three types of MBL-associated serine proteases, MASP-1, MASP-2 and MASP-3, and a truncated form of MASP-2 (small MBL-associated protein; sMAP or MAp19) complex with MBL to activate the lectin pathway of the complement system. Activated MASPs subsequently cleave and activate downstream components of the complement pathway. MASP-3 is an alternatively spliced product from the MASP-1 gene and may function to inhibit MASP-2 by competing for MBL binding and inhibiting the activation of MBLassociated MASP-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MASP2
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MASP2
Gene Accession No. O00187
Gene Alias mannan binding lectin serine peptidase 2; mannan-binding; mannan-binding lectin serine peptidase 1 pseudogene 1; mannan-binding lectin serine peptidase 2; mannan-binding lectin serine peptidase 2 variant 1; mannan-binding lectin serine peptidase 2 variant 2; mannan-binding lectin serine peptidase 2 variant 3; mannan-binding lectin serine peptidase 2 variant 4; mannan-binding lectin serine protease 1 pseudogene 1; mannan-binding lectin serine protease 2; Mannan-binding lectin serine protease 2 A chain; Mannan-binding lectin serine protease 2 B chain; mannose binding lectin-associated serine protease-2; mannose-binding protein-associated serine protease 2; MAp19; MASP1P1; Masp2; MASP-2; MBL-associated plasma protein of 19 kD; MBL-associated serine protease 2; sb:eu714; small MBL-associated protein; sMAP; wu:fb62a09
Gene Symbols MASP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MASP2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10747
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.