Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Maspin Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1767R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1767R7 7 mL Prediluted
MS1767S1 500 μL Concentrated
MS1767S 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1767R7 Supplier Epredia™ Supplier No. MS1767R7

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ Maspin Ab-1, Mouse Monoclonal Antibody.

Maspin or mammary-specific serpin is a tumor suppressor protein of serine proteinase inhibitor family. Its transcription is regulated in combinatorial fashion by Ets, p53 and AP-1. It is present in normal breast and prostatic epithelial cells but downregulated in the respective carcinomas. The extent of downregulation correlates with the progression of the carcinoma and may help in prognosis.

Host Species: Mouse

Clone: EAW24

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein corresponding to N-terminal region of human Maspin

Molecular Weight: 42kDa

Positive Control: Prostate

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Maspin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EAW24
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminal region of human Maspin
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.