Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Maspin Polyclonal Antibody, Invitrogen™

Catalog No. PIPA589591
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589591 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589591 Supplier Thermo Scientific Supplier No. PA589591

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Rabbit Polyclonal Antibody

Immunogen sequence: EKEPLGNVLF SPICLSTSLS LAQVGAKGDT ANEIGQVLHF ENVKDVPFGF QTVTSDVNKL SSFYSLKLIK RLYVDKSLNL STEFISSTKR PYAKELETVD FKDKLEETKG QINNSIKDLT DGHFENILAD NSVNDQTKIL VVNAAYFVGK WMKKFSESET KECPFRVNK; Positive Samples: HT-29, NIH/3T3; Cellular Location: extracellular space

SERPINB5 is a tumor suppressor. It blocks the growth, invasion, and metastatic properties of mammary tumors. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, it exhibits no serine protease inhibitory activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Maspin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene SERPINB5
Gene Accession No. P36952, P70124
Gene Alias 1110036M19Rik, AI462524, AI646751, Maspin, ovalbumin, peptidase inhibitor 5, PI5, PI-5, protease inhibitor 5, protease inhibitor 5 (maspin), serine (or cysteine) peptidase inhibitor, clade B, member 5, serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 5, serine (or cysteine) proteinase inhibitor, clade B, member 5, serine protease inhibitor 7, serpin B5, serpin family B member 5, serpin peptidase inhibitor, clade B (ovalbumin), member 5, SERPINB5, Spi5, Spi7
Gene Symbols SERPINB5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 21-189 of human SERPINB5 (NP_0026302)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20724, 5268
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.