Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation Mass Spec UPLC Column Kit

Catalog No. 50791065 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
Diameter (Metric):
2.1 mm
3 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
50-791-065 100 mm 3 mm
50-791-022 100 mm 2.1 mm
50-791-021 50 mm 2.1 mm
50-791-064 50 mm 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-791-065 Supplier Waters Corporation Supplier No. 176001888
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The MS UPLC method development kit combines BEH C18, with HSS C18, C18 SB and T3 column chemistries to accomodate your method development approach, enabling methods to be developed efficiently and effectively.

  • By utilizing UPLC Technology for method development, a substantial improvement in throughput can be realized.
  • This, in turn, reduces cost per sample and time of analysis considerably while maintaining or improving separation integrity, enabling products to be brought to market faster and improving the overall profitability of the assay.

Specifications

Diameter (Metric) 3 mm
Length (Metric) 100 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 1.7 μm
Pore Size 130 Å
Quantity 4/Pk.
Endcapped No
Product Type Column Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.