Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mast Cell Chymase Monoclonal Antibody (CC1), Invitrogen™

Catalog No. PIMA511717
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511717 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511717 Supplier Thermo Scientific Supplier No. MA511717

Mouse Monoclonal Antibody

MA5-11717 targets Mast Cell Chymase in IHC (P) and WB applications and shows reactivity with Human samples. The MA5-11717 immunogen is purified human skin chymase.

Mast Cell Chymase Monoclonal antibody specifically detects Mast Cell Chymase in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mast Cell Chymase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CC1
Concentration 80 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene CMA1
Gene Accession No. P23946
Gene Alias alpha-chymase; Chymase; chymase 1; chymase 1 preproprotein transcript E; chymase 1 preproprotein transcript I; chymase 1, mast cell; chymase, heart; chymase, mast cell; CMA1; CYH; CYM; mast cell; Mast cell chymase 1; mast cell protease 3; Mast cell protease 5; mast cell protease I; Mast cell protease III; MCP3P; Mcp5; Mcp-5; Mcpt3; Mcpt5; MCT1; mMCP-5; rMCP-3; rMCP-5; rMCP-III
Gene Symbols CMA1
Host Species Mouse
Immunogen Purified human skin chymase
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1215
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.