Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mast Cell Marker Antibody (MCG35), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250484FR
Description
Mast Cell Marker Monoclonal specifically detects Mast Cell Marker in Human samples. It is validated for Immunohistochemistry.Specifications
Mast Cell Marker | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
Spleen cells and bone marrow cells (erythrocyte depleted) from a patient with systemic mastocytosis. The bone marrow preparation consisted of 50% typical mast cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Immunohistochemistry | |
MCG35 | |
Immunohistochemistry | |
Mouse | |
Protein A purified | |
Cardiovascular Biology | |
The antibody does not react with basophils or other haemopoietic cell types. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction