Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mast Cell Protease-1/Mcpt1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298243JF549
Description
Mast Cell Protease-1/Mcpt1 Polyclonal antibody specifically detects Mast Cell Protease-1/Mcpt1 in Mouse samples. It is validated for ELISASpecifications
Mast Cell Protease-1/Mcpt1 | |
Polyclonal | |
ELISA | |
AV080368, Mcp-1 | |
Produced in rabbits immunized with purified, recombinant Mouse Mast Cell Protease-1/Mcpt1 (Accession#: NP_032596.1; Met 1-Lys 246) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
17224 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction