Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568011
Description
Predicted to react with Mouse and Rat samples.
Appears to link the dystrophin/utrophin network with microtubule filaments via the syntrophins. Phosphorylation of DMD or UTRN may modulate their affinities for associated proteins.
Specifications
| MAST1 | |
| Polyclonal | |
| Unconjugated | |
| MAST1 | |
| 9430008B02Rik; KIAA0973; MAST1; mast1 {ECO:0000312; microtubule associated serine/threonine kinase 1; microtubule-associated serine/threonine-protein kinase 1; RGD:631372}; SAST; SAST170; syntrophin associated serine/threonine kinase; syntrophin-associated serine/threonine-protein kinase; syntrophin-associated serine-threonine protein kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22983, 353118, 56527 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q810W7, Q9R1L5, Q9Y2H9 | |
| MAST1 | |
| Synthesized peptide derived from human MAST1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction